Learn More
Cayman Chemical 5-hydroxy Tryptophol 10mg
SDP
Supplier:  Cayman Chemical 1611410 MG
Encompass_Preferred
A metabolite of tryptophan that is formed by the alcohol dehydrogenase-catalyzed reduction of the serotonin intermediate, 5-hydroxyindoleacetaldehyde; used as a biomarker for recent alcohol consumption
Catalog No. 50-296-8785