Learn More
Medchemexpress LLC 5-Hydroxyindole-3-acetic acid | 54-16-0 | 191.19 | 500 MG

Supplier: Medchemexpress LLC HYW008253500MG
5-Hydroxyindole-3-acetic acid is the main metabolite of serotonin or metanephrines, and it can be used as a biomarker of neuroendocrine tumors. It is for research use only.
- Formula: C10H9NO3
- Appearance: Solid
- Color: Yellow to brown
- Purity: 99.45%
- Molecular weight: 191.19
- Stores at 4°C under nitrogen. In solvent, stores at -80°C for 6 months; -20°C for 1 month, under nitrogen.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.