Learn More
Medchemexpress LLC 5-Hydroxyindole-3-acetic acid-d2 | 56209-31-5 | 193.20 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW008253S1MG
Encompass_Preferred
5-Hydroxyindole-3-acetic acid-d2 is the deuterium-labeled form of 5-Hydroxyindole-3-acetic acid. 5-Hydroxyindole-3-acetic acid is the main metabolite of serotonin or metanephrines and can be used as a biomarker for neuroendocrine tumors. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-47882