Learn More
Apexbio Technology LLC 5-Hydroxymethyl-2'-deoxyuidine-5-monophosphate, disodium salt 5238-86-8(fee acid) 10mg
SDP
Supplier:  Apexbio Technology LLC B892010
Encompass_Preferred
5-Hydroxymethyl-2p-deoxyuidine-5-monophosphate disodium salt is a modified nucleotide derivative involved in DNA methylation and demethylation pathways It incorporates a hydroxymethyl group at the 5-position of the pyrimidine ring of deoxyuridine monophosphate Enzymes such as DNA methyltransferases and demethylases utilize this compound as a substrate or intermediate during DNA epigenetic regulation Based on these properties 5-Hydroxymethyl-2p-deoxyuidine-5-monophosphate disodium salt holds research potential in studies of DNA repair replication fidelity epigenetic reprogramming genomic stability enzymatic activity assays and epigenome profiling
Catalog No. 50-000-89416