Learn More
Medchemexpress LLC 5-Hydroxymethyl-2'-deoxyuridine | 5116-24-5 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12998310MM/1ML
Encompass_Preferred
5-Hydroxymethyl-2'-deoxyuridine is a nucleoside analog that inhibits the replication of multiple human leukemia cell lines. It is suitable for cell replication and leukemia research.
  • Inhibits replication of multiple human leukemia cell lines
  • Prolongs survival in mice with L1210 leukemia
  • Suitable for cell replication and leukemia research
  • Exhibits dose-dependent toxicity against acute promyelocytic leukemia cell lines
  • Available in solid form or as a pre-prepared solution
Catalog No. 50-003-38549