Learn More
Medchemexpress LLC 5-Hydroxymethyl-2'-deoxyuridine | 5116-24-5 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY129983
Encompass_Preferred
5-Hydroxymethyl-2'-deoxyuridine is a nucleoside analog that inhibits the replication of various human leukemia cell lines with IC50 values between 1.7 and 5.8 μM. It has also been shown to prolong the survival of mice with L1210 leukemia. This compound is suitable for research concerning cell replication and leukemia. It has a molecular weight of 258.23 and a chemical formula of C10H14N2O6, appearing as a white to off-white solid.
  • Inhibits replication of multiple human leukemia cell lines
  • Prolongs survival of mice carrying L1210 leukemia
  • Inhibits replication of Sarcoma 180 cells and Ehrlich ascites carcinoma cells
  • Inhibits herpes simplex virus type 1 (HSV-1) pyrimidine 2'-deoxyribonucleoside kinase
  • Shows dose-dependent toxicity against human acute promyelocytic leukemia cell line
Catalog No. 50-003-38783