Learn More
Medchemexpress LLC 5-Hydroxymethyl-2-furancarboxylic acid | 6338-41-6 | 142.11 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00524110MM/1ML
Encompass_Preferred
5-Hydroxymethyl-2-furancarboxylic acid is the main metabolite of 5-hydroxymethyl-2-furfural (HMF) in the body and eliminated renally. It can be used as a biomarker for the exposure to HMF. This product is for research use only.
  • Main metabolite of 5-hydroxymethyl-2-furfural (HMF)
  • Eliminated renally
  • Can be used as a biomarker for HMF exposure
  • Appears as a white to yellow solid
  • Soluble in DMSO (100 mg/mL)
  • Functions as a microbial and human endogenous metabolite
Catalog No. 50-004-49571