Learn More
Medchemexpress LLC 5-(hydroxymethyl)cytidine | 19235-17-7 | MFCD26523159 | ≥99.0% | 273.24 | C10H15N3O6 | 10 MG

Supplier: Medchemexpress LLC HY15285410MG
5-(Hydroxymethyl)cytidine is a cytidine nucleoside analogue used as a research reagent to study DNA and RNA modifications, cell-cycle effects, and nucleoside metabolism. Supplied as a small-quantity solid for preparing stock solutions and in vitro assays.
- High purity (≥99.0%).
- Molecular formula C10H15N3O6; molecular weight 273.24.
- CAS number 19235-17-7.
- Soluble in water (~20.83 mg/mL); sonication may be required.
- Store at -20°C as a sealed solid; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
- Used to probe DNA damage and cell-cycle pathways as a nucleoside antimetabolite/analog.
- Available in 1 mg, 5 mg, and 10 mg quantities for preparing stock solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.