Learn More
AdipoGen 5-Hydroxynicotinic acid
SDP
Supplier:  AdipoGen CDXH0052G005
Encompass_Preferred
Chemical. CAS 27828-71-3. Formula C6H5NO3. MW 139.11. Synthetic A nicotinic acid analog. Substrate analog MHPCO, used in the investigation of the hydroxylation mechanism of the flaovoprotein 2-methyl-3-hydroxypyridine-5-carboxylic acid oxygenase MHPCO. Also used as building block for synthesis. Hydroxynicotinic acid is formed by the enzymatic action of nicotinate with nicotinic acid hydroxylase or nicotinate hydroxylase.
Catalog No. 50-256-0777