Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Hydroxypyrazine-2-Carboxylic Acid | 34604-60-9 | 99.99% | C5H4N2O3 | 5 G
SDP

Supplier:  Medchemexpress LLC HY762105G

Encompass_Preferred

5-Hydroxypyrazine-2-Carboxylic Acid is a metabolite of the anti-tuberculosis drug pyrazinamide (PZA). It is intended for research use only and has a high purity of 99.99%.

  • Metabolite of anti-tuberculosis drug pyrazinamide (PZA)
  • For research use only
  • High purity of 99.99%
  • Solid appearance
  • Molecular weight: 140.10
  • Molecular formula: C5H4N2O3
  • CAS number: 34604-60-9
  • Off-white to light brown in color
  • Consistent 1H NMR spectrum
  • Soluble in DMSO and water

Catalog No. 50-004-34681


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.