Learn More
eMolecules​ 5-HYDROXYQUINOLINE-3-CARBOXYLIC ACID | AstaTech | 911108-90-2 | MFCD18417133 | 189.170 | C10H7NO3 | 95.000 | OC(=O)c1cnc2cccc(O)c2c1 | 0.25g | 200618596
Supplier:  eMolecules​ 200618596
Encompass_Preferred
5-HYDROXYQUINOLINE-3-CARBOXYLIC ACID | AstaTech | 911108-90-2 | MFCD18417133 | 189.170 | C10H7NO3 | 95.000 | OC(=O)c1cnc2cccc(O)c2c1 | 0.25g | 200618596
Catalog No. NC2046865