Learn More
Medchemexpress LLC 5-Hydroxyquinoline | 578-67-6 | 99.5% | 145.16 | 100 G

Supplier: Medchemexpress LLC HY32100100G
5-Hydroxyquinoline is an organic compound commonly used in certain chemical reactions and biological research. It can be used in the manufacture of dyes, cellulase, oxidants and passivators, and is widely applied in the pigment, paint, and rubber industries. Additionally, it serves as a precursor, intermediate, and metal ion complexing agent for certain drugs.
- Appearance: Solid
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month
- Solubility: In DMSO at 50 mg/mL (344.45 mM)
- Usage: For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.