Learn More
Medchemexpress LLC 5-Hydroxyquinoline | 578-67-6 | 99.5% | 145.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY32100
Encompass_Preferred
5-Hydroxyquinoline is an organic compound used in various chemical reactions and biological research. It finds application in the manufacture of dyes, cellulase, oxidants, and passivators, and is extensively utilized in the pigment, paint, and rubber industries. Additionally, it serves as a precursor, intermediate, and metal ion complexing agent for certain drugs, and is a biochemical reagent for life science-related research.
  • Used in the manufacture of dyes, cellulase, oxidants, and passivators
  • Extensively utilized in the pigment, paint, and rubber industries
  • Serves as a precursor, intermediate, and metal ion complexing agent for drugs
  • Biochemical reagent for life science-related research
Catalog No. 50-004-02897