Learn More
Medchemexpress LLC HY-N0122 10mg Medchemexpress, 5-Hydroxytryptophan CAS:56-69-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN012210MG
Encompass_Preferred
Medchemexpress, HY-N0122 10mg 5-Hydroxytryptophan CAS:56-69-9 5-Hydroxytryptophan, a tryptophan metabolite, is a direct 5-hydroxytryptamine (5-HT) precursor and an L-aromatic amino acid decarboxylase substrate. [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3409