Learn More
Medchemexpress LLC HY-W041019 10mg Medchemexpress, 5-Hydroxytryptophol CAS:154-02-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW04101910MG
Encompass_Preferred
Medchemexpress, HY-W041019 10mg 5-Hydroxytryptophol CAS:154-02-9 5-Hydroxytryptophol is a mammalian serotonin metabolite, acting as a marker of acute alcohol consumption. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4658