Learn More
Medchemexpress LLC 5-iodopicolinic acid | 32046-43-8 | MFCD09910324 | 249.01 | C6H4INO2 | 5 G

Supplier: Medchemexpress LLC HYW0416575G
5-iodopicolinic acid (5-iodopyridine-2-carboxylic acid) is an iodinated pyridine carboxylic acid used primarily as an intermediate in organic synthesis for preparing other active compounds.
- Used as an intermediate in organic synthesis
- Iodinated pyridine carboxylic acid with aromatic iodine substituent
- Molecular formula C6H4INO2, molecular weight 249.01
- CAS number 32046-43-8
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.