Learn More
Medchemexpress LLC 5-iodopyridine-2-carboxylic acid | 32046-43-8 | MFCD09910324 | 249.01 | C6H4INO2 | 500 MG

Supplier: Medchemexpress LLC HYW041657500MG
5-Iodopicolinic acid (5-iodopyridine-2-carboxylic acid) is an iodine-containing pyridine carboxylic acid used as an intermediate in organic synthesis. It is supplied as a solid and is typically used for building block and synthetic transformations.
- used as an intermediate in organic synthesis.
- iapac name: 5-iodopyridine-2-carboxylic acid.
- molecular formula C6H4INO2, molecular weight 249.01 g/mol.
- cas number 32046-43-8 for unambiguous identification.
- available in multiple pack sizes, including 500 mg and 1 g; larger sizes available by inquiry.
- store at 4°C and protect from light; in solvent, store at -80°C (6 months) or -20°C (1 month).
- batch-specific purity and analytical data are provided in the CoA and purity documentation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.