Learn More
Selleck Chemical LLC 5-Iodotubercidin 5mg 24386-93-4 NSC 113939, 5-ITu
SDP
Supplier:  Selleck Chemical LLC S83145MG
Encompass_Preferred
5-Iodotubercidin (NSC 113939, 5-ITu) is a potent adenosine kinase inhibitor with IC50 of 26 nM. It inhibits nucleoside transporter, CK1, insulin receptor tyrosine kinase, phosphorylase kinase, PKA, CK2 and PKC. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5934