Learn More
Medchemexpress LLC HY-15424 5mg , 5-Iodotubercidin CAS:24386-93-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY154245MG
Encompass_Preferred
HY-15424 5mg 5-Iodotubercidin CAS: 24386-93-4 5-Iodotubercidin (NSC 113939), an ATP mimetic, is a potent adenosine kinase inhibitor with an IC50 of 26 nM. 5-Iodotubercidin (NSC 113939) initiates glycogen synthesis in isolated hepatocytes by causing inactivation of phosphorylase and activation of glycogen synthase. 5-Iodotubercidin (NSC 113939) also inhibits CK1, insulin receptor tyrosine kinase, phosphorylase kinase, PKA, CK2, PKC and Haspin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9366