Learn More
Cayman Chemical 5-IodourIdIn 5g
SDP
Supplier:  Cayman Chemical 356975 G
Encompass_Preferred
A pyrimidine nucleoside derivative; inhibits dihydroorotase (Ki = 340 µM for the rat enzyme); sensitizes Don Chinese hamster lung cells to irradiation; has been used in the synthesis of compounds with antiviral and anticancer activities
Catalog No. 50-291-1389