Learn More
Medchemexpress LLC 5-Iodouridine | 1024-99-3 | 99.9% | 370.10 | 1 G

Supplier: Medchemexpress LLC HYW0110791G
5-Iodouridine is an iodine-containing pyrimidine nucleoside analog. It inhibits dihydroorotase with a Ki value of 340 μM. It significantly enhances the cell-killing effect of gamma irradiation and can be used in the research of HSV-1 infection and leukemia.
- Iodine-containing pyrimidine nucleoside analog
- Inhibits dihydroorotase
- Enhances cell-killing effect of gamma irradiation
- Suitable for research of HSV-1 infection
- Suitable for research of leukemia
- High purity (99.93%)
- White to off-white solid appearance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.