Learn More
Medchemexpress LLC 5-Iodouridine | 1024-99-3 | 99.9% | 370.10 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01107910G
Encompass_Preferred
5-Iodouridine is an iodine-containing pyrimidine nucleoside analog that inhibits dihydroorotase. It significantly enhances the cell-killing effect of gamma irradiation and is utilized in research for various conditions.
  • Iodine-containing pyrimidine nucleoside analog
  • Inhibits dihydroorotase with a Ki value of 340 μM
  • Significantly enhances the cell-killing effect of gamma irradiation
  • Useful in research for HSV-1 infection
  • Useful in research for leukemia
  • Used in research for indolent lymphoid malignancies
  • Exhibits antitumor activity
Catalog No. 50-004-52011