Learn More
Cayman Chemical 5-IPF2a-VI 100ug
SDP
Supplier:  Cayman Chemical 16300100 UG
Encompass_Preferred
A potent inhibitor of both human and mouse soluble epoxide hydrolase (IC50 3.7 and 2.8 nM respectively) has superior pharmacokinetics compared to the 1-adamantylurea based inhibitors like AUDA
Catalog No. 50-297-2629