Learn More
Cayman Chemical 5-IPF2a-VI 50ug
SDP
Supplier:  Cayman Chemical 1630050 UG
Encompass_Preferred
A water-soluble derivative of geldanamycin which potently inhibits Hsp90 (IC50 24 nM) and has excellent bioavailability and tissue distribution in animals has diverse anti-tumor actions suppresses inflammation by interfering with signaling through the NF-KB pathway ameliorates high fat diet-induced renal failure
Catalog No. 50-297-2042