Learn More
Medchemexpress LLC 5-Isoxazoleacetic acid, α-amino-2,3-dihydro-3-oxo- | 2552-55-8 | MFCD00069294 | 1 ML

Supplier: Medchemexpress LLC HYN231110MM/1ML
Ibotenic acid is a potent iGluR agonist, exhibiting activity at both N-methyl-D-aspartate (NMDA) and metabolotropic quisqualate (Qm) receptor sites. This psychoactive compound, naturally occurring in certain mushroom species, is a conformationally restricted analog of L-Glutamic acid. It is neurotoxic to cortical neurons with an EC50 of 77.3±8 μM. With a purity of 98.83% (HPLC), it is intended for research use only. Storage recommendations are -80°C for 2 years or -20°C for 1 year when in solvent.
- Agonist for NMDA and Qm receptor sites
- Neurotoxic activity in cortical neurons
- Conformationally restricted analog of L-Glutamic acid
- Purity of 98.83% (HPLC)
- Storage recommendations: -80°C for 2 years, -20°C for 1 year (in solvent)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.