Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences 5-Layer Tissue Culture Flask, Plug Seal Cap, Sterile, Treated - 870 cm² per surface, 1/pk (8/cs)
SDP

Supplier:  Chemglass Life Sciences CGN2404V02

Encompass_Preferred

Multi-layer 5-layer tissue culture flask designed for high-surface-area cell growth applications. Constructed from high-clarity, 100% virgin polystyrene with vacuum plasma tissue culture treatment and E-beam sterilization, each flask provides five stacked growth surfaces to increase culture capacity while maintaining lot traceability and individual wrapping.

  • High clarity, 100% virgin polystyrene.
  • Sterilized by E-beam.
  • Vacuum plasma tissue culture treatment.
  • Growth area per surface: 870 cm².
  • Five stacked growth surfaces (5-layer).
  • Plug seal cap with straight neck.
  • Pack size: 1 per pack, 8 per case.
  • Individually plastic wrapped.
  • Non-pyrogenic and DNase/RNase-free.
  • Clear lot number for batch traceability.

Catalog No. 50-273-3439


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.