Learn More
Medchemexpress LLC 5-Methoxy-2-benzimidazolethiol | 37052-78-1 | 180.23 | 10 MM 1 ML

Supplier: Medchemexpress LLC HYW00837810MM/1ML
5-Methoxy-2-benzimidazolethiol is a benzimidazole derivative extensively studied for its chemical properties, including its iodimetric determination in alkaline media. This compound appears as a solid, ranging from white to light yellow, and is available in solution format for various research applications.
- High purity of 99.6%
- Molecular weight of 180.23
- Chemical formula: C8H8N2OS
- CAS number: 37052-78-1
- Appearance: Solid, white to light yellow
- Soluble in DMSO at 100 mg/mL
- Powder storage: -20°C for 3 years or 4°C for 2 years
- Solution storage: -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.