Learn More
Medchemexpress LLC 5-Methoxy-2-benzimidazolethiol | 37052-78-1 | 180.23 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00837850G
Encompass_Preferred
5-Methoxy-2-benzimidazolethiol is a benzimidazole compound used for research purposes. Studies have investigated its iodimetric determination in alkaline media.
  • Solid appearance
  • White to light yellow in color
  • Soluble in DMSO at 100 mg/mL
  • Stable as a powder for 3 years at -20°C and 2 years at 4°C
  • Stable in solvent for 6 months at -80°C and 1 month at -20°C
Catalog No. 50-004-47435