Learn More
Medchemexpress LLC 5-methoxy-5-oxopentanoic acid | 1501-27-5 | MFCD00004409 | 99.3% | 146.14 g/mol | C6H10O4 | 10 G

Supplier: Medchemexpress LLC HYW01752310G
5-Methoxy-5-oxopentanoic acid is an endogenous metabolite offered as a high-purity analytical standard for research, metabolomics, and analytical method development. The product is supplied in multiple pack sizes to accommodate small-scale assays and larger preparative needs, and is characterized with specified purity and physical property data for use as a reference material.
- Endogenous metabolite suitable for metabolomics studies.
- High purity: 99.28%.
- Molecular formula C6H10O4; molecular weight 146.14 g/mol.
- Available in multiple pack sizes, including a 10 g option, for experimental flexibility.
- Colorless to light yellow liquid with density 1.164 g/cm3.
- Intended for use as an analytical reference standard and for method development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.