Learn More
Apexbio Technology LLC 5-Methoxy-CTP(Synonyms: 5-Methoxycytidine triphosphate, 5-Methoxy-CTP, 5-MeO-CTP, 5-Methoxy Cytidine-5'-triphosphate), 10x100ul (100 mM).
SDP
Supplier:  Apexbio Technology LLC B80621
Encompass_Preferred
5-Methoxy-CTP is a chemically modified cytidine triphosphate analog, characterized by a methoxy group substitution at position 5 of the cytosine ring. During in vitro transcription, it can be enzymatically incorporated into synthesized RNA, resulting in RNAs containing 5-methoxy-modified cytidine residues. This methoxy substitution influences RNA chemical properties, alters its structural characteristics, and enhances resistance against intracellular nucleolytic degradation. Consequently, 5-Methoxy-CTP is valuable for investigating RNA structural dynamics, RNA-protein interactions, and RNA functional roles, and it holds potential utility in RNA-based therapeutics, functional genomics research, and gene therapy approaches.
Catalog No. 50-000-60712