Learn More
Selleck Chemical LLC 5-Methoxytryptamine
SDP
Supplier:  Selleck Chemical LLC S632425MG
Encompass_Preferred
5-Methoxytryptamine (Mexamine Methoxytryptamine) is a tryptamine derivative that acts as a full agonist at the 5-HT1 5-HT2 5-HT4 5-HT6 and 5-HT7 receptors
Catalog No. 50-000-03861