Learn More
Medchemexpress LLC 5-Methoxytryptamine | 608-07-1 | MFCD00005662 | 99.9% | 190.25 g/mol | C11H14N2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01516910MM/1ML
Encompass_Preferred
5-Methoxytryptamine (O-Methylserotonin) is an indoleamine serotonin (5-HT) receptor agonist supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is formulated for in vitro pharmacology and biochemical research, including receptor binding and signaling studies.
  • Ready-to-use 10 mM solution in DMSO for immediate assay use.
  • High purity (99.91%) suitable for pharmacological studies.
  • Consistent concentration supports reproducible results.
  • Appropriate for receptor binding and signaling assays.
  • Small-volume format reduces freeze-thaw and waste.
Catalog No. 50-003-03372