Learn More
Matrix Scientific 5-METHYL-2-PYRIDIN-3-YLM 500MG
Supplier:  Matrix Scientific 019430500MG
Encompass_Preferred
5-METHYL-2-PYRIDIN-3-YLMETHYL-2H-PYRAZOL-3-YLAMINE 100mg CAS#957500-08-2 MFCD07186471 C10H12N4 MW:188.23
Catalog No. NC2222700