Learn More
Cayman Chemical 5-methyl-7-MethoxyIsoflavon 5g
SDP
Supplier:  Cayman Chemical 392675 G
Encompass_Preferred
A synthetic isoflavone; inhibits the clonogenic survival of HCT116 cells at 10, 20, and 40 µM
Catalog No. 50-291-6578