Learn More
Medchemexpress LLC 5-Methylcytidine | 2140-61-6 | 257.24 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY113135R10MG
Encompass_Preferred
5-Methylcytidine is a nucleoside compound that exhibits antiviral activity, with an IC50 value of 0.06 μM against HSV-1. This compound is formed by the attachment of a methyl group to the 5th carbon atom of cytosine. It is a common modification found in DNA, playing a significant role in processes such as gene expression regulation and development, and is closely related to the occurrence and development of various diseases, including cancer.
  • Exhibits antiviral activity against HSV-1.
  • Plays a significant role in gene expression regulation and development.
  • Related to the occurrence and development of various diseases.
Catalog No. 50-004-35756