Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methylindole | 614-96-0 | 131.17 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW00197210G

Encompass_Preferred

5-Methylindole is a biochemical reagent suitable for life science related research. It appears as a white to off-white solid.

  • Appearance: White to off-white solid
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • Solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in DMSO: 100 mg/mL

Catalog No. 50-004-47343


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.