Learn More
Medchemexpress LLC 5-Methylpyrazine-2-carboxylic acid | 5521-55-1 | 99.9% | 138.12 | 100 G
Supplier: Medchemexpress LLC HY32739100G
5-Methylpyrazine-2-carboxylic acid is a biochemical reagent suitable for life science research applications. This solid compound is for research use only and not intended for patient use. It requires careful handling, as it may cause serious eye damage.
- Designed for research applications
- Solid appearance
- Requires protective measures during handling
- Store in a well-ventilated area, away from direct sunlight
- Can be dissolved in DMSO, PEG300, Tween-80, or corn oil for in vivo studies