Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methylsalicylic acid | 89-56-5 | 99.8% | C8H8O3 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY7857310MM/1ML

Encompass_Preferred

5-Methylsalicylic acid, also known as p-Cresotic acid, is a biochemical reagent supplied as a 10 mM solution in DMSO. This high-purity compound is intended for life science research and development use only, and not for medicinal, household, or other applications.

  • Purity: 99.8%
  • Chemical formula: C8H8O3
  • Molecular weight: 152.15
  • Exhibits an IC50 of 9.8 mM against HEK293 cells
  • Store in solvent at -80°C for up to 2 years or -20°C for up to 1 year

Catalog No. 50-004-34442


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.