Learn More
Medchemexpress LLC 5-Methyltetrahydrofolic acid | 134-35-0 | 98.6% | 459.46 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113046R100MG
Encompass_Preferred
This compound is an endogenous metabolite and a folate metabolic intermediate. It serves as an analytical standard, commonly utilized as a reference standard in qualitative, quantitative, and methodological research experiments, including those involving HPLC, GC, and MS.
  • Application: used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Structure classification: ketones, aldehydes, acids.
  • Initial source: endogenous metabolite, human gut microbiota metabolites.
  • Related classifications: natural products, endogenous metabolite, microorganisms, human gut microbiota metabolites, food-sourced standards, vitamin standards.
Catalog No. 50-004-39753