Learn More
Medchemexpress LLC 5-Methyltetrahydrofolic acid | 134-35-0 | 99.77% | 459.46 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11304610MM/1ML
Encompass_Preferred
5-Methyltetrahydrofolic acid (5-Methyl THF) is the main circulating form of folic acid in the body and is involved in a variety of biochemical reactions. It regulates cardiovascular function by increasing the production of endothelin-1 (ET-1) in low-density lipoprotein-treated endothelial cells and can be used in the study of cardiovascular diseases.
  • Main circulating form of folic acid in the body.
  • Involved in a variety of biochemical reactions.
  • Regulates cardiovascular function by increasing endothelin-1 (ET-1) production.
  • Can be used in the study of cardiovascular diseases.
  • Increases the production of ET-1 in endothelial cells in a concentration-dependent manner (0, 10, 50 and 100 μM; 24 h).
  • Inhibits the production of thiobarbituric acid reactive substances (TBARS) by HUVEC in the presence of low-density lipoprotein (100 μM; 24 h).
Catalog No. 50-004-39778