Learn More
Medchemexpress LLC 5-Methyltetrahydrofolic acid (standard) | 134-35-0 | 98.6% | 459.46 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY113046R50MG
Encompass_Preferred
The compound is an analytical standard, which serves as a reference standard assay. It is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for reference assays.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • Reference standard for food testing, natural products, and vitamins.
  • Endogenous metabolite and human gut microbiota metabolite.
  • Related to ketones, aldehydes, and acids.
Catalog No. 50-004-39732