Learn More
Andwin Scientific 5 ML PP STORAGE VIAL WITH SCRE
SDP
Supplier:  Andwin Scientific 15051510
Encompass_Preferred
The 5 mL self-standing screw cap tube offer convenience and security with a self-standing skirt and an HDPE screw cap with a molded sealing ring. It also has a large, etched writing surface that allow easy sample identification and graduations for precise volume measurement. It comes in 20 bags of 25 vials, totaling to 500 vials.
Catalog No. 50-819-831