Learn More
Medchemexpress LLC HY-103173 10mg Medchemexpress, 5'-N-Ethylcarboxamidoadenosine CAS:35920-39-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10317310MG
Encompass_Preferred
Medchemexpress, HY-103173 10mg 5'-N-Ethylcarboxamidoadenosine CAS:35920-39-9 5'-N-Ethylcarboxamidoadenosine (NECA) is a nonselective adenosine receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8049