Learn More
Medchemexpress LLC 5-NIdR 5mg | 191421-10-0 | 5 MG

Supplier: Medchemexpress LLC HY1155675MG
5-NIdR is a research-grade nucleoside analog (1-(β-D-2-deoxyribofuranosyl)-5-nitroindole) supplied as a light yellow solid for laboratory use. It has reported high purity and is commonly used in cell-based assays investigating DNA/RNA processes and apoptosis.
- High purity (98.0%).
- Available as solid (5 mg, 10 mg, 25 mg) and solution (10 mM in DMSO).
- Molecular weight 278.26 g/mol; formula C13H14N2O5.
- Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).
- Intended for in vitro research and cell-based assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.