Learn More
Medchemexpress LLC 5-nitro-2-(3-phenylpropylamino)benzoic acid | 107254-86-4 | MFCD00153851 | >=98.0% | 300.31 g/mol | C16H16N2O4 | 5 MG

Supplier: Medchemexpress LLC HY1010125MG
NPPB (5-nitro-2-(3-phenylpropylamino)benzoic acid) is a small-molecule research reagent used to inhibit outwardly rectifying chloride channels (ORCC) in in vitro studies. It is supplied as a high-purity solid suitable for analytical and biochemical assays.
- Blocks outwardly rectifying chloride channels (ORCC).
- Chemical name: 5-nitro-2-(3-phenylpropylamino)benzoic acid.
- CAS number: 107254-86-4.
- Molecular weight: 300.31 g/mol.
- Chemical formula: C16H16N2O4.
- Purity: ≥98%.
- Supplied as a 5 MG quantity for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.