Learn More
Medchemexpress LLC 5-nitro-2-{[5-(phenoxymethyl)-4-phenyl-4H-1,2,4-triazol-3-yl]thio}pyridine | 345989-24-4 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12152325MG
Encompass_Preferred
MIND4-17 is a small-molecule NRF2 activator that covalently modifies the C151 residue of Keap1, disrupting Keap1-Nrf2 association and stabilizing Nrf2 to induce antioxidant response element (ARE) gene expression. Supplied as a solid with high reported purity, it is used in cellular and in vivo studies of oxidative stress pathways.
  • Activates Nrf2 signaling by modifying Keap1 C151.
  • Promotes expression of ARE-regulated antioxidant genes such as Nqo1 and Hmox1.
  • Reported molecular weight 405.43 g/mol and formula C20H15N5O3S.
  • Purity reported as 99.93% on the product page.
  • Supplied as an off-white to light yellow solid in multiple sizes, including 25 mg.
Catalog No. 50-002-32386