Learn More
Medchemexpress LLC 5-nitro-2-furaldehyde diacetate | 92-55-7 | MFCD00003244 | 100g
SDP
Supplier:  Medchemexpress LLC HYY1826100G
Encompass_Preferred
IITR01324 is an antimicrobial agent IITR01324 has antimicrobial activity against Escherichia coli (MIC 3 58 mg/L) and other Gram-negative bacteria (Shigella flexneri MIC 3 58 mg/L Cronobacter sakazakii MIC 28 63 mg/L) IITR01324 exerts its antimicrobial activity by destroying bacterial DNA after activation of intracellular reductases IITR01324 can be used to study the development of new antimicrobial drugs especially against multidrug-resistant (MDR) pathogens[1]
Catalog No. 50-002-66337