Learn More
Medchemexpress LLC 5-nitro-6-(3-nitrophenyl)-2-oxo-4-(trifluoromethyl)-1H-pyridine-3-carbonitrile | 685121-45-3 | 99.9% | 354.20 g·mol⁻1 | C13H5F3N4O5 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1248761MG
Encompass_Preferred
SynuClean-D is a small-molecule inhibitor of α-synuclein aggregation used in research to study amyloid fibril formation and neurodegeneration. It disrupts mature fibrils, prevents fibril propagation, and has shown neuroprotective effects in animal models of Parkinson's disease.
  • Inhibits α-synuclein aggregation and disrupts mature fibrils.
  • Prevents fibril propagation and protects dopaminergic neurons in animal models.
  • High purity suitable for research use (99.9%).
  • Molecular weight 354.20 g·mol⁻1; chemical formula C13H5F3N4O5.
  • Powder storage: -20°C for long-term; in solvent: -80°C recommended for extended storage.
  • Readily soluble in DMSO (~83.33 mg/mL); in vivo formulations yield clear solutions ≥2.08 mg/mL.
Catalog No. 50-002-38831