Learn More
Medchemexpress LLC 5-nitro-6-(3-nitrophenyl)-2-oxo-4-(trifluoromethyl)-1H-pyridine-3-carbonitrile | 685121-45-3 | MFCD04111032; MFCD31735118; MFCD32015709 | 99.9% | 354.20 g/mol | C13H5F3N4O5 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12487610MM 1ML
Encompass_Preferred
SynuClean-D is a small-molecule inhibitor of α-synuclein aggregation used in research to disrupt amyloid fibrils and inhibit fibril propagation; it has shown neuroprotective effects in Parkinson's disease models.
  • Inhibits α-synuclein aggregation in vitro.
  • Disrupts mature amyloid fibrils.
  • Prevents fibril propagation and seeding.
  • Reduces dopaminergic neuron degeneration in animal models.
  • Available as a 10 mM solution in DMSO and in multiple solid quantities.
  • High assay purity (≈99.9%).
  • Molecular formula C13H5F3N4O5, molecular weight 354.20 g/mol.
Catalog No. 50-002-39322