Learn More
Medchemexpress LLC 5-Nitrobenzimidazole | 94-52-0 | 99.7% | 163.14 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW010132100G
Encompass_Preferred
5-nitrobenzimidazole is a genotoxic nitro compound and a 6-aminopurine analogue. It exhibits a slight inhibitory effect on xanthine oxidase, with an IC50 of 86.84 μM. This compound can be utilized in hyperuricemia research.
  • Genotoxic nitro compound
  • 6-aminopurine analogue
  • Slight inhibitory effect on xanthine oxidase
  • IC50: 86.84 μM (xanthine oxidase)
  • Can be used in research of hyperuricemia
  • Appearance: Solid
  • Color: White to yellow
Catalog No. 50-004-53584